- Pk:100 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:1286
- Protein synonyms:24p3|MSFI|NGAL|p25
- Protein/peptide name:Lipocalin-2
- Purity:≥94%
Lipocalin-2 (LCN2) is a member of the lipocalin superfamily, which share a highly conserved fold containing an eight-stranded antiparallel beta barrel (Flower), and transport small hydrophobic molecules. Playing a role in innate immunity, Lipocalin-2 acts as a potent bacteriostatic reagent by sequestering iron-containing siderophores (Flo et al.). It is mainly expressed in activated neutrophils, but is also secreted by other immune cells in response to infection. Lipocalin-2 has been implicated in multiple cellular processes, such as differentiation (Yang et al.), migration (Du et al.), and apoptosis (Devireddy et al.). In vitro studies suggest that lipocalin-2 is a potential therapeutic target for certain cancers, including colorectal cancer (CRC; Zhang et al.) and chronic lymphocytic leukemia (Bauvois et al.). This protein product contains a His-residue tag at the carboxyl end of the polypeptide chain. For consistency and reproducibility across your applications, lipocalin-2 from STEMCELL comes lyophilised with ≥94% purity, and endotoxin levels are verified to be ≤1.0 EU/μg protein.