Order Entry
Austria
ContactUsLinkComponent
Lipopolysaccharide from E. coli (O55:B5)
Lipopolysaccharide from E. coli (O55:B5)
Catalog # STMC100-1270
CAS Number:  
undefined
Lipopolysaccharide from E. coli (O55:B5)
Catalog # STMC100-1270
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Seq:
    2070
  • Protein/peptide name:
    Lipopolysaccharide from E. coli (O55:B5)
  • Purity:
    Ultrapure(≥99.9%). NodetectableDNA,RNA,orproteintraces.

Specifications

About this item

Trigger a variety of immunological responses with E. coli Lipopolysaccharide O55:B5 (S-form), a lipopolysaccharide (LPS) derived from the O55:B5 serotype of the Gram-negative bacteria and nbsp Escherichia coli. Composed of a lipid A, a core oligosaccharide, and an O antigen, LPS are glycolipid constituents that reside on the outer membranes of gram-negative bacteria (Kitchens RL et al.). LPS protects bacteria against bile salts and lipophilic antibiotics by maintaining the outer integrity of the cell membrane (Bäckhed F et al.). E. coli lipopolysaccharide O55:B5 (S-form), in particular, is predominantly recognized by toll-like receptor 4 (TLR4), which leads to the activation of NF-κβ, a protein complex which plays a key role in regulating immune response (Kuzmich N et al.). Activation of NF-κβ can trigger increased production of pro-inflammatory cytokines IL-1 and TNF-α by macrophages (Matuschak GM et al.). This LPS can also interact with CD14 to activate phospholipase Cγ2 and kinases of the Src family, trigger influxes of extracellular Ca2+, as well as calcineurin-dependent translocation of the nuclear factor of activated T cells (NFAT) family of transcription factors (Li CC et al.). When added to ImmunoCult™-SF macrophage medium (Catalog #10961), stimulation with lipopolysaccharide from E. coli (O55:B5) and IFN-γ supports the polarization to M1 (classically activated) macrophages. Warning: This product is highly pyrogenic. Avoid all means by which the product may enter the bloodstream.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').