Nitrogen (Total) Standard Solution, CRM
Catalog # CMPLTN0100.L1
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
CAS Number:
Specifications
- Concentration:100 mg/L N
- Pk:100 mL
- Description:Nitrogen (total) standard solution
Specifications
About this item
Total nitrogen standard (100 mg/L) - for photometric applications.
Certifications: EN 25663 (ISO 5663) and ISO 7150. ISO 17034, ISO 17025, ISO 9001.
Ordering information: Shelf life on ship date: 24 Months - Store under normal laboratory conditions, at temperatures between 15 to 25 °C.