Intracellular Permeabilisation Buffer (10X)
Catalog # STMC100-1451
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:150 mL
- Description:Reagent for the permeabilisation of fixed cells
Specifications
About this item
Ideal for the permeabilisation of fixed cells during labeling of cytokines and other cytoplasmic antigens. Intracellular Permeabilisation Buffer (10X) is uniquely formulated for optimal resolution and to reduce background and is suitable for use with fluorochrome-conjugated antibodies for flow cytometry.
Application: Immunology
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').