Dulbecco's Phosphate Buffered Saline (DPBS), with 2% Fetal Bovine Serum
Catalog # STMC07905
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:500 mL
- Description:Cell culture buffer
Specifications
About this item
Dulbecco's Phosphate Buffered Saline with 2% fetal bovine serum is recommended for use as a buffer to dilute or wash nucleated cells.
Species: Human, mouse, non human primate, other, rat
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').