Order Entry
Austria
ContactUsLinkComponent
Anti-Adherence Rinsing Solution
Catalog # STMC07010
undefined
Anti-Adherence Rinsing Solution
Catalog # STMC07010

Specifications

  • Description:
    Rinsing solution for cultureware to prevent cell adhesion
  • Pk:
    100 mL

Specifications

About this item

Anti-Adherence Rinsing Solution is a surfactant solution for pre-treating cultureware to reduce surface tension and prevent cell adhesion. It can be used with plates, flasks, pipettes, strainers and other cultureware.

Adherence rinsing solution is required for use with the following:
- AggreWell™ plates (e.g. Catalog MSPP-34811)
- Organoid growth media, including HepatiCult™ (Catalog MSPP-06030)
- PancreaCult™ (Catalog MSPP-06040)

Application: Cancer, neuroscience, stem cell biology
Species: Human, mouse, non human primate, other, rat

Caution: Note: formerly known as AggreWell™ Rinsing Solution, name change effective Dec 2017.
Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').