Order Entry
Austria
Orders LinkContactUsLinkComponent
RoboSep™ Buffer 2

RoboSep™ Buffer 2

Buffer containing BSA, 250 mL
Catalog # STMC20164
Marketsource
RoboSep™ Buffer 2
Catalog # STMC20164
Marketsource
CAS Number:  
RoboSep™ Buffer 2
RoboSep™ Buffer 2

Buffer containing BSA, 250 mL

CAS Number:  
Price
94,10 €
Log in for availability
1 * 250 mL
Product Details & Documents

RoboSep™ buffer 2 is recommended for select EasySep™ cell separation protocols.

Application: Immunology
Species: Human, mouse, non human primate, other, rat

Please note that one or two bottles of buffer are included with every purchase of a RoboSep™ reagent kit.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Buffer containing BSA

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.