Order Entry
Austria
ContactUsLinkComponent
EasySep™ Buffer
EasySep™ Buffer
Catalog # STMC20144
undefined
EasySep™ Buffer
Catalog # STMC20144

Specifications

  • Description:
    Cell separation buffer
  • Pk:
    1 L

Specifications

About this item

EasySep™ buffer is recommended for EasySep™ cell separation protocols.

Application: Chimerism, Hla, immunology
Species: Human, mouse, non human primate, other, rat

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').