Order Entry
Austria
ContactUsLinkComponent
Human Recombinant Flt3/Flk-2 Ligand (E. coli expressed)
Human Recombinant Flt3/Flk-2 Ligand (E. coli expressed)
Certificates
undefined
Human Recombinant Flt3/Flk-2 Ligand (E. coli expressed)

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein synonyms:
    FL|FLT3L|Flt3-L|Flt3 Ligand|Fms-like tyrosine kinase 3 ligand
  • Protein/peptide name:
    Flt3/Flk-2 Ligand
  • Purity:
    ≥95%

Specifications

Frequently Bought Together