Order Entry
Austria
Orders LinkContactUsLinkComponent
Human Recombinant GM-CSF (CHO-expressed) | Avantor

Human Recombinant GM-CSF (CHO-expressed)

Marketsource
Human Recombinant GM-CSF (CHO-expressed)
Marketsource
Certificates
Human Recombinant GM-CSF (CHO-expressed)

3 Options Available Below

Product Details & Documents

Granulocyte-macrophage colony-stimulating factor (GM-CSF) promotes the proliferation and differentiation of hematopoietic progenitor cells and the generation of neutrophils, eosinophils, and macrophages. In synergy with other cytokines such as stem cell factor, IL-3, erythropoietin, and thrombopoietin, it also stimulates erythroid and megakaryocyte progenitor cells (Barreda et al.). GM-CSF is produced by multiple cell types, including stromal cells, Paneth cells, macrophages, dendritic cells (DCs), endothelial cells, smooth muscle cells, fibroblasts, chondrocytes, and Th1 and Th17 T cells (Francisco-Cruz et al.). The receptor for GM-CSF (GM-CSFR) is composed of two subunits: the cytokine-specific α subunit (GMRα; CD116) and the common subunit βc (CD131) shared with IL-3 and IL-5 receptors (Broughton et al.). GM-CSFR is expressed on hematopoietic cells, including progenitor cells and immune cells, as well as non-hematopoietic cells. Recombinant human GM-CSF (rhGM-CSF) promotes the production of myeloid cells of the granulocytic (neutrophils, eosinophils, and basophils) and monocytic lineages in vivo. It has been tested for mobilisation of hematopoietic progenitor cells and used to treat chemotherapy-induced neutropenia in patients. GM-CSF is able to stimulate the development of DCs that ingest, process, and present antigens to the immune system (Francisco-Cruz et al.).

Achtung! Bitte beachten: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • STMC78190.2
  • STMC78190
  • STMC78190.1
  • Protein/peptide name:
  • GM-CSF
  • GM-CSF
  • GM-CSF
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Human
  • Source:
  • CHO cells
  • CHO cells
  • CHO cells
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 1.000 µG
    Log in for availability
    Log in for availability
    1 * 1.000 µG
    1 * 1.000 µG - 6 370,00 €
    6 370,00 €
    1 * 1.000 µG
       6 370,00 €

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC78190.2
    Protein/peptide name
    GM-CSF
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    CHO cells
    Purity
    ≥95%
    Marketsource
  • 10 µG
    Log in for availability
    Log in for availability
    1 * 10 µG
    1 * 10 µG - 308,00 €
    308,00 €
    1 * 10 µG
       308,00 €

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC78190
    Protein/peptide name
    GM-CSF
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    CHO cells
    Purity
    ≥95%
    Marketsource
  • 50 µG
    Log in for availability
    Log in for availability
    1 * 50 µG
    1 * 50 µG - 617,00 €
    617,00 €
    1 * 50 µG
       617,00 €

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC78190.1
    Protein/peptide name
    GM-CSF
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    CHO cells
    Purity
    ≥95%
    Marketsource
Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen.