Order Entry
Austria
ContactUsLinkComponent
Human Recombinant SCF, ACF
Human Recombinant SCF, ACF
Certificates
undefined
Human Recombinant SCF, ACF

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    SCF, ACF
  • Purity:
    ≥95%

Specifications

Frequently Bought Together