Order Entry
Austria
Orders LinkContactUsLinkComponent
Human Recombinant IL-6, ACF

Human Recombinant IL-6, ACF

Marketsource
Human Recombinant IL-6, ACF
Marketsource
Certificates
Human Recombinant IL-6, ACF

3 Options Available Below

Product Details & Documents

Interleukin 6 (IL-6) is a pleiotropic growth factor with the wide range of biological activities in immune regulation, hematopoiesis, and oncogenesis. IL-6 is produced by a variety of cell types including T cells, B cells, monocytes and macrophages, fibroblasts, hepatocytes, vascular endothelial cells, and various tumor cell lines. On its own or in combination with other factors such as IL-2 and interferon-γ, IL-6 stimulates the proliferation of B cells, T cells, and hybridoma cells (Hirano et al.; Mihara et al.; Tanaka et al). In combination with cytokines such as IL-3, GM-CSF and SCF, IL-6 has been shown to promote hematopoietic progenitor cell proliferation and differentiation in vitro. IL-6 signals through a cell surface type I cytokine receptor complex consisting of the ligand-binding IL-6α (CD126) and the signal-transducing gp130 subunits. The binding of IL-6 to its receptor system includes activation of JAK/STAT signaling pathway (Mihara et al.; Peters et al.; Tanaka et al.). This product is animal component-free.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • STMC78148.2
  • STMC78148
  • STMC78148.1
  • Protein/peptide name:
  • IL-6, ACF
  • IL-6, ACF
  • IL-6, ACF
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Human
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 1.000 µG
    Log in for availability
    Log in for availability
    1 * 1.000 µG
    1 * 1.000 µG - 4 930,00 €
    4 930,00 €
    1 * 1.000 µG
       4 930,00 €

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC78148.2
    Protein/peptide name
    IL-6, ACF
    Protein/peptide type
    Recombinant
    Species
    Human
    Purity
    ≥95%
    Marketsource
  • 20 µG
    Log in for availability
    Log in for availability
    1 * 20 µG
    1 * 20 µG - 308,00 €
    308,00 €
    1 * 20 µG
       308,00 €

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC78148
    Protein/peptide name
    IL-6, ACF
    Protein/peptide type
    Recombinant
    Species
    Human
    Purity
    ≥95%
    Marketsource
  • 100 µG
    Log in for availability
    Log in for availability
    1 * 100 µG
    1 * 100 µG - 1 030,00 €
    1 030,00 €
    1 * 100 µG
       1 030,00 €

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC78148.1
    Protein/peptide name
    IL-6, ACF
    Protein/peptide type
    Recombinant
    Species
    Human
    Purity
    ≥95%
    Marketsource
Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen.