Mouse Recombinant IL-21
About this item
Interleukin 21 (IL-21) is a pleiotropic cytokine that is composed of four α-helical bundles and primarily produced by natural killer T (NKT) cells, T follicular helper (Tfh) cells, and Th17 cells (Spolski and Leonard 2008). IL-21 signals via receptor heterodimerization of IL-21 receptor and IL-2 receptor subunit gamma (IL-2RG or CD132), both of which have a common gamma-chain subunit and activate the JAK/STAT, MAPK, and PI3K pathways (Parrish-Novak et al.; Ozaki et al. 2000; Spolski and Leonard 2014). IL-21 has been shown to have a critical role in regulating immunoglobulin production and differentiation of the pro-inflammatory Th17 population of cells (Ozaki et al. 2002; Nurieva et al.). Additionally, IL-21 specifically sustains CD8+ T cell effector activity and provides a mechanism of CD4+ T cell help during chronic viral infection (Elsaesser et al.). IL-21 signaling was also found critical for the development of type 1 diabetes in non-obese diabetic (NOD) mice (Sutherland et al.) and control of T cell autoimmunity by regulatory B cells (Yoshizaki et al.).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
3 Options Available Below
Product Details & Documents
Interleukin 21 (IL-21) is a pleiotropic cytokine that is composed of four α-helical bundles and primarily produced by natural killer T (NKT) cells, T follicular helper (Tfh) cells, and Th17 cells (Spolski and Leonard 2008). IL-21 signals via receptor heterodimerization of IL-21 receptor and IL-2 receptor subunit gamma (IL-2RG or CD132), both of which have a common gamma-chain subunit and activate the JAK/STAT, MAPK, and PI3K pathways (Parrish-Novak et al.; Ozaki et al. 2000; Spolski and Leonard 2014). IL-21 has been shown to have a critical role in regulating immunoglobulin production and differentiation of the pro-inflammatory Th17 population of cells (Ozaki et al. 2002; Nurieva et al.). Additionally, IL-21 specifically sustains CD8+ T cell effector activity and provides a mechanism of CD4+ T cell help during chronic viral infection (Elsaesser et al.). IL-21 signaling was also found critical for the development of type 1 diabetes in non-obese diabetic (NOD) mice (Sutherland et al.) and control of T cell autoimmunity by regulatory B cells (Yoshizaki et al.).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- STMC78116.2
- STMC78116
- STMC78116.1
- Protein/peptide name:
- IL-21
- IL-21
- IL-21
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Species:
- Mouse
- Mouse
- Mouse
- Purity:
- ≥95%
- ≥95%
- ≥95%
Specifications
Product Family Options
Product Information
- PkAvailabilityPrice
- Specifications:Marketsource
More
Specifications
Cat. No.STMC78116.2Protein/peptide nameIL-21Protein/peptide typeRecombinantSpeciesMousePurity≥95%Marketsource - Specifications:Marketsource
More
Specifications
Cat. No.STMC78116Protein/peptide nameIL-21Protein/peptide typeRecombinantSpeciesMousePurity≥95%Marketsource - Specifications:Marketsource
More
Specifications
Cat. No.STMC78116.1Protein/peptide nameIL-21Protein/peptide typeRecombinantSpeciesMousePurity≥95%Marketsource



