Order Entry
Austria
Orders LinkContactUsLinkComponent
Human Recombinant IL-3 (<i>E. coli </i>expressed)

Human Recombinant IL-3 (E. coli expressed)

Marketsource
Human Recombinant IL-3 (<i>E. coli </i>expressed)
Marketsource
Certificates
Human Recombinant IL-3 (<i>E. coli </i>expressed)

3 Options Available Below

Product Details & Documents

Interleukin 3 (IL-3) is a species-specific pleiotropic cytokine that promotes the survival and proliferation of pluripotent hematopoietic stem cells and lineage-committed progenitor cells and their differentiation into mature cells of most lineages, including basophils, neutrophils, eosinophils, macrophages, dendritic cells, erythrocytes, and megakaryocytes (Yang et al.; Dorssers et al.; Broughton et al.). IL-3 is produced by activated T cells and has a physiological role in inflammation and allergies by promoting the secretion of inflammatory mediators such as histamine, IL-4, and IL-6 by basophils and eosinophils (Broughton et al.). The IL-3 receptor consists of a unique alpha subunit (CD123) and a beta common subunit (βc or CD131) that is shared with the receptors for IL-5 and GM-CSF, and is the principal signal transduction subunit for these cytokines. IL-3 binding to the heterodimeric receptor activates JAK/STAT, MAPK, and PI3K signaling pathways (Woodcock et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • STMC78040.1
  • STMC78040
  • STMC78040.2
  • Protein/peptide name:
  • IL-3
  • IL-3
  • IL-3
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 10 µG
    Log in for availability
    Log in for availability
    1 * 10 µG
    1 * 10 µG - 275,00 €
    275,00 €
    1 * 10 µG
       275,00 €

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC78040.1
    Protein/peptide name
    IL-3
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    E. coli
    Purity
    ≥95%
    Marketsource
  • 100 µG
    Log in for availability
    Log in for availability
    1 * 100 µG
    1 * 100 µG - 987,00 €
    987,00 €
    1 * 100 µG
       987,00 €

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC78040
    Protein/peptide name
    IL-3
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    E. coli
    Purity
    ≥95%
    Marketsource
  • 1.000 µG
    Log in for availability
    Log in for availability
    1 * 1.000 µG
    1 * 1.000 µG - 4 160,00 €
    4 160,00 €
    1 * 1.000 µG
       4 160,00 €

    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    STMC78040.2
    Protein/peptide name
    IL-3
    Protein/peptide type
    Recombinant
    Species
    Human
    Source
    E. coli
    Purity
    ≥95%
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.