Human Recombinant IGF-II
Supplier: STEMCELL Technologies
Certificates
About this item
Insulin-like growth factor 2 (IGF-II) is a polypeptide that belongs to the family of insulin-like growth factors that are similar in molecular structure to proinsulin. IGF-II binding to IGF-I receptor activates a signaling cascade via the MAPK and PI3K pathways. IGF-II affects growth, differentiation, and survival of cells in a variety of tissues during embryonic development. It stimulates proliferation and migration of primary lymphatic endothelial cells, and induces lymphangiogenesis (Guvakova).
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
- Protein/peptide type:Recombinant
- Species:Human
- Protein synonyms:Insulin-like growth factor II|Insulin-like growth factor type 2|Somatomedin A|T3M-11-derived growth factor
- Protein/peptide name:IGF-II
- Purity:≥95%
Specifications
Frequently Bought Together