Order Entry
Austria
ContactUsLinkComponent
 
Mouse Recombinant IFN-gamma
 
undefined
Mouse Recombinant IFN-gamma

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Protein/peptide name:
    IFN-gamma
  • Purity:
    ≥95%

 

Frequently Bought Together