Order Entry
Austria
ContactUsLinkComponent
Mouse Recombinant IL-19
Mouse Recombinant IL-19
Certificates
undefined
Mouse Recombinant IL-19

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Protein/peptide name:
    IL-19
  • Purity:
    ≥95%

Specifications

Frequently Bought Together