Interleukin 3 (IL-3) is a species-specific pleiotropic cytokine that promotes the survival and proliferation of pluripotent hematopoietic stem cells and lineage-committed progenitor cells and their differentiation into mature cells of most lineages, including basophils, neutrophils, eosinophils, macrophages, dendritic cells, erythrocytes, and megakaryocytes (Yang et al.; Dorssers et al.; Broughton et al.). IL-3 is produced by activated T cells and has a physiological role in inflammation and allergies by promoting the secretion of inflammatory mediators such as histamine, IL-4, and IL-6 by basophils and eosinophils (Broughton et al.). The IL-3 receptor consists of a unique alpha subunit (CD123) and a beta common subunit (βc or CD131) that is shared with the receptors for IL-5 and GM-CSF, and is the principal signal transduction subunit for these cytokines. IL-3 binding to the heterodimeric receptor activates JAK/STAT, MAPK, and PI3K signaling pathways (Woodcock et al.).
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Protein/peptide name:IL-3
- Purity:≥95%