Order Entry
Austria
ContactUsLinkComponent
 
Human Recombinant HBEGF
 
undefined
Human Recombinant HBEGF

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    HBEGF
  • Purity:
    ≥95%

 

Frequently Bought Together