Order Entry
Austria
ContactUsLinkComponent
Human Recombinant NT-4
Human Recombinant NT-4
Certificates
undefined
Human Recombinant NT-4

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    NT-4
  • Purity:
    ≥95%

Specifications

Frequently Bought Together