Order Entry
Austria
ContactUsLinkComponent
 
Human Recombinant MCP-1 (CCL2)
 
undefined
Human Recombinant MCP-1 (CCL2)

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    MCP-1 (CCL2)
  • Purity:
    ≥95%

 

Frequently Bought Together