- Pk:100 µG
- Protein/peptide type:Recombinant
- Species:Human
- Protein synonyms:NAMPT|PBEF|PBEF1
- Protein/peptide name:Visfatin
- Purity:≥90%
Use visfatin to catalyse the production of nicotinamide mononucleotide from nicotinamide (Revollo et al.). Nicotinamide mononucleotide is a precursor to nicotinamide adenine dinucleotide (NAD) that is vital to energy metabolism, cell death, and other cellular processes (Ying). Visfatin also acts as an immunomodulator by activating leukocytes and inducing the production of pro-inflammatory and anti-inflammatory cytokines (Moschen et al.), and there is evidence that visfatin can regulate insulin receptor signaling and insulin secretion (Brown et al.). Belonging to the nicotinate phosphoribosyltransferase (NAPRTase) family, Visfatin is an adipokine that is produced in adipocytes, leukocytes, and hepatocytes (Chiu et al., Garten et al., Kralisch et al.). It is active as a homodimer with each identical monomer consisting of two structural domains made up of 19 β-strands and 13 α-helices (Kim et al.). For consistency and reproducibility across your applications, visfatin from STEMCELL comes lyophilised with ≥90% purity, and is verified by LAL analysis to ensure endotoxin levels are ≤1,0 EU/μg protein.