Human Recombinant Ikaros (IKZF1) (NM_006060)
Catalog # ORIGTP313207M
Supplier: OriGene
CAS Number:
Specifications
- Pk:100 µl
- Protein/peptide type:Recombinant
- Species:Human
- Tag sequence:C-Myc/DDK
- Storage conditions:25 mM Tris-HCl, 100 mM glycine, pH 7,3, 10% glycerol, Store at –80 °C
- Protein synonyms:PRO0758|ZNFN1A1|LyF-1|PPP1R92|CVID13|LYF1|IK1|IKAROS|Hs.54452
- UniProtKB:Q13422, R9R4D9
- Protein/peptide name:Ikaros (IKZF1)
- Purity:>80% as determined by SDS-PAGE and Coomassie blue staining
- Molecular weight:57,3 kDa
- Concentration:>0,05 µg/µl as determined by microplate BCA method
Specifications
About this item
This gene encodes a transcription factor that belongs to the family of zinc-finger DNA-binding proteins associated with chromatin remodeling. The expression of this protein is restricted to the fetal and adult hemo-lymphopoietic system, and it functions as a regulator of lymphocyte differentiation.