ContactUsLinkComponent
SLK Human siRNA Oligo Duplex (Locus ID 9748)
SLK Human siRNA Oligo Duplex (Locus ID 9748)
Catalog # ORIGSR306513
Supplier:  OriGene
New Product
CAS Number:  
SLK Human siRNA Oligo Duplex (Locus ID 9748)
Catalog # ORIGSR306513
Supplier:  OriGene
New Product
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    SLK Human siRNA oligo duplex (Locus ID 9748)
Specifications
About this item
This SLK Human siRNA Oligo Duplex mediates apoptosis and actin stress fiber dissolution.
  • HPLC purified
  • Tested by ESI-MS
Can be used for approximately 330 transfections/2 nmol in 24-well plate under optimised conditions (final conc. 10 nM).
Delivery: Kit contains SLK (Human) - 3 unique 27mer siRNA duplexes - 2 nmol each (Locus ID 9748), Trilencer-27 universal scrambled negative control siRNA duplex - 2 nmol, RNAse free siRNA duplex resuspension buffer - 2 ml.