ContactUsLinkComponent
ReproSil-Pur® Basic C8 HPLC Columns
ReproSil-Pur® Basic C8 HPLC Columns
Supplier:  DR MAISCH
New Product
Certificates
ReproSil-Pur® Basic C8 HPLC Columns
Supplier:  DR MAISCH
New Product
Specifications
  • Catalog No:
  • MAISR15.B8.S1546
  • MAISR15.B9.S0404
  • Carbon load:
  • 15,0
  • 17,0
  • Endcapped:
  • Yes
  • Yes
  • pH range:
  • 2 - 9
  • 2 - 9
  • Surface area:
  • 450 m²/g
  • 450 m²/g
  • USP code:
  • L7
  • L1