Order Entry
Austria
ContactUsLinkComponent
Supelco® Discovery® BIO Reversed-Phase Solution to Protein and Peptide Separation Challenges
Supelco® Discovery® BIO Reversed-Phase Solution to Protein and Peptide Separation Challenges
Supplier:  Merck
Certificates
Supelco® Discovery® BIO Reversed-Phase Solution to Protein and Peptide Separation Challenges
Supplier:  Merck

Specifications

  • Catalog No:
  • 567225-U
  • 568323-U
  • Application:
  • Peptide mapping/proteolytic digests/small biomolecules
  • Peptide mapping/proteolytic digests/small biomolecules
  • Carbon load:
  • 5,0%
  • 5,0%
  • Endcapped:
  • Yes
  • Yes
  • Max. pressure:
  • 400 bar
  • 400 bar
  • pH range:
  • 2 - 8
  • 2 - 8
  • Surface area:
  • 100 m²/g
  • 100 m²/g
  • Temperature limit:
  • 0...70 °C
  • 0...70 °C
  • Shipping conditions:
  • Acetonitril/water
  • Acetonitril/water

Specifications

Frequently Bought Together