Cutan® Wall Dispenser for Hand Sanitiser
Catalog # EVONCPROB01SA
Supplier: Deb-STOKO
New Product
Specifications
- Capacity:1 L
- Description:Wall dispenser
- Environmentally Preferable:
- Pk:1
Specifications
About this item
Cutan dispenser for use with any Cutan 1 Litre hand sanitiser cartridge.
- Contains silver phosphate glass technology to prevent microbial growth
- Strong and durable
- Key operated lock prevents theft
- Large push button that can be hand or elbow operated
- Easy to slot in and replace the cartridge
Red coloured label and push button for easy distinction and identification of hand sanitiser amongst hand cleaning and moisturising agents. No special DIY skills required, can be easily mounted with the sticky pad at the back of the dispenser. Unit self-primes on the first stroke of the button.