Order Entry
Austria
ContactUsLinkComponent
Supelco® Ascentis® Silica HPLC Columns
Supelco® Ascentis® Silica HPLC Columns
Supplier:  Merck
Certificates
Supelco® Ascentis® Silica HPLC Columns
Supplier:  Merck

Specifications

  • Catalog No:
  • 581515-U
  • 581510-U
  • 581519-U
  • 581509-U
  • 581518-U
  • 581514-U
  • 581513-U
  • 581503-U
  • 581502-U
  • 581512-U
  • 581522-U
  • 581500-U
  • Application:
  • Small molecule separation
    Food and beverage
    HPLC and LC/MS suitable
  • Small molecule separation
    Food and beverage
    HPLC and LC/MS suitable
  • Small molecule separation
    Food and beverage
    HPLC and LC/MS suitable
  • Small molecule separation
    Food and beverage
    HPLC and LC/MS suitable
  • Small molecule separation
    Food and beverage
    HPLC and LC/MS suitable
  • Small molecule separation
    Food and beverage
    HPLC and LC/MS suitable
  • Small molecule separation
    Food and beverage
    HPLC and LC/MS suitable
  • Small molecule separation
    Food and beverage
    HPLC and LC/MS suitable
  • Small molecule separation
    Food and beverage
    HPLC and LC/MS suitable
  • Small molecule separation
    Food and beverage
    HPLC and LC/MS suitable
  • Small molecule separation
    Food and beverage
    HPLC and LC/MS suitable
  • Small molecule separation
    Food and beverage
    HPLC and LC/MS suitable
  • Max. pressure:
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • pH range:
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • Surface area:
  • 450 m²/g
  • 450 m²/g
  • 450 m²/g
  • 450 m²/g
  • 450 m²/g
  • 450 m²/g
  • 450 m²/g
  • 450 m²/g
  • 450 m²/g
  • 450 m²/g
  • 450 m²/g
  • 450 m²/g
  • Temperature limit:
  • ≤70 °C
  • ≤70 °C
  • ≤70 °C
  • ≤70 °C
  • ≤70 °C
  • ≤70 °C
  • ≤70 °C
  • ≤70 °C
  • ≤70 °C
  • ≤70 °C
  • ≤70 °C
  • ≤70 °C
  • USP code:
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • Shipping conditions:
  • Ethanol
  • Ethanol
  • Ethanol
  • Ethanol
  • Ethanol
  • Ethanol
  • Ethanol
  • Ethanol
  • Ethanol
  • Ethanol
  • Ethanol
  • Ethanol

Specifications

Related Information

Frequently Bought Together