Michigan Pesticide Mixture 2
Catalog # EHERA50000100AL
Supplier: EHRENSTORFER
New Product
CAS Number:
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pk:1 mL
- Description:Michigan pesticide mixture 2
- Storage conditions:−18 °C
- GHS signal:Danger
- Storage temperature:20 °C
- Form:Mixture