Order Entry
Austria
ContactUsLinkComponent
Human Recombinant HGF (from Insect cells)
Human Recombinant HGF (from Insect cells)
Catalog # ENZORLT-300-010
Supplier:  Enzo Life Sciences
CAS Number:  
Human Recombinant HGF (from Insect cells)
Catalog # ENZORLT-300-010
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    5 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Storage conditions:
    After reconstitution, prepare aliquots and store at −20 °C. Avoid freeze/thaw cycles. Short term storage: Ambient. Long term storage: −20 °C.
  • Endotoxin content:
    ≤1,2 EU/µg protein
  • Biological activity:
    ED50=0,5 - 1,0 ng/ml (Scattering activity in MDCK cell assays)
  • Reconstitution instructions:
    Reconstitute with water (100 µg/ml)
  • Protein synonyms:
    Hepatocyte growth factor|HGF (human) (recombinant)|SF (human) (recombinant)|Scatter factor (human) (recombinant)
  • Protein/peptide name:
    HGF
  • Purity:
    ≥90% (SDS-PAGE)
  • Solubility:
    Soluble in water and most aqueous buffers
  • Molecular weight:
    ~78 kDa
  • Formulation:
    Lyophilised
  • Shipping temperature:
    Ambient temperature

Specifications

About this item

Caution: Research use only.