Bacterial identification, Diatabs
Catalog # ROSC48811
Supplier: ROSCO
CAS Number:
Specifications
- Pk:250
- Description:Tributyrin
Specifications
About this item
Diatabs offers an accurate and rapid identification of clinically relevant bacteria and yeasts. The concept of individual tests - used as single tests or in combination - offers the user many advantages over traditional methods. The Diatabs system offers the opportunity of finding the most appropriate and economical tests for a number of purposes.
- Rapid (4 hour) enzymatic and modified conventional test
- Detection of natural susceptibility patterns or growth factor requirement
- An extensive range of more than 80 individual tests