Order Entry
Austria
Orders LinkContactUsLinkComponent
 

pH-Fix pH Indicator Test Strips

pH-Fix pH Indicator Test Strips
Supplier:  MACHEREY-NAGEL
Certificates
pH-Fix pH Indicator Test Strips

17 Options Available Below

Product Details & Documents

pH-Fix indicator test strips allow fast pH testing directly at the point of interest, and are suitable for untrained users as well as analytical professionals.

  • No accessories needed - always ready to use
  • Brilliant colour chart for precise readings
  • Long handle for sufficient hand - sample - distance
  • Non bleeding - no contamination of sample solutions

The indicator dyes used for pH-Fix test strips are chemically bound to the test pads to prevent bleeding and, therefore, contamination of the sample. Instructions for use according to IVD Directive 98/79/EC - The instructions for use according to IVD Directive 98/79/EC can be requested by providing the article number and LOT number at the email address [email protected].

 : No hazardous material.

Specifications for Product Family

Specifications

  • Catalog No:
  • 662-0208
  • 662-0210
  • 662-2337
  • 662-2343
  • 662-2344
  • 662-0200
  • 662-2341
  • 662-0201
  • 662-2342
  • 662-0240
  • 662-2338
  • 662-0241
  • 662-0242
  • 662-2339
  • 662-0207
  • 662-0236
  • 662-0237
  • Shelf life:
  • 4 Year(s)
  • 4 Year(s)
  • 4 Year(s)
  • 4 Year(s)
  • 4 Year(s)
  • 4 Year(s)
  • 4 Year(s)
  • 4 Year(s)
  • 4 Year(s)
  • 4 Year(s)
  • 4 Year(s)
  • 4 Year(s)
  • 4 Year(s)
  • 4 Year(s)
  • 4 Year(s)
  • 4 Year(s)
  • 4 Year(s)

Specifications

Product Family Options

Classic flat box container with colour chart

Product Information

  • pH range
    Pack type
    Availability
    Price
  • 662-0208
    pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax
    pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax
    0,0 - 6,0
    Classic flat box , 65×90×10 mm, 21 g
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,10 €
    16,10 €
    1 * 100 ST
       16,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    662-0208
    Shelf life
    4 Year(s)
  • 662-0210
    pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax
    pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax
    0 - 14
    Classic flat box , 66×10×90 mm, 35,1 g
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,10 €
    16,10 €
    1 * 100 ST
       16,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    662-0210
    Shelf life
    4 Year(s)
  • 662-2343
    pH-Fix, 100 test strips with colour scale
    pH-Fix, 100 test strips with colour scale
    0,3-2,3
    Classic flat box , 65×10×90 mm, 32,4 g
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,10 €
    16,10 €
    1 * 100 ST
       16,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    662-2343
    Shelf life
    4 Year(s)
  • 662-2344
    pH-Fix, 100 test strips with colour scale
    pH-Fix, 100 test strips with colour scale
    1,7 - 3,8
    Classic flat box , 66×90×12 mm, 36 g
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,10 €
    16,10 €
    1 * 100 ST
       16,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    662-2344
    Shelf life
    4 Year(s)
  • 662-0200
    pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax
    pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax
    2,0 - 9,0
    Classic flat box , 66×10×90 mm, 34,95 g
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,10 €
    16,10 €
    1 * 100 ST
       16,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    662-0200
    Shelf life
    4 Year(s)
  • 662-2341
    pH-Fix, 100 test strips with colour scale
    pH-Fix, 100 test strips with colour scale
    3,1 - 8,3
    Classic flat box , 66×10×90 mm, 34,43 g
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,10 €
    16,10 €
    1 * 100 ST
       16,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    662-2341
    Shelf life
    4 Year(s)
  • 662-0201
    pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax
    pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax
    3,6 - 6,1
    Classic flat box , 61×90×10 mm, 20 g
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,10 €
    16,10 €
    1 * 100 ST
       16,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    662-0201
    Shelf life
    4 Year(s)
  • 662-2342
    pH-Fix, 100 test strips with colour scale
    pH-Fix, 100 test strips with colour scale
    4,0 - 7,0
    Classic flat box , 68×90×15 mm, 32 g
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,10 €
    16,10 €
    1 * 100 ST
       16,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    662-2342
    Shelf life
    4 Year(s)
  • 662-0240
    pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax
    pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax
    4,5 - 10,0
    Classic flat box , 66×10×90 mm, 35,61 g
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,10 €
    16,10 €
    1 * 100 ST
       16,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    662-0240
    Shelf life
    4 Year(s)
  • 662-0241
    pH-Fix, 100 test strips with colour scale
    pH-Fix, 100 test strips with colour scale
    5,1 - 7,2
    Classic flat box , 65×10×90 mm, 35,9 g
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,10 €
    16,10 €
    1 * 100 ST
       16,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    662-0241
    Shelf life
    4 Year(s)
  • 662-0242
    pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax
    pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax
    6,0 - 7,7
    Classic flat box , 65×10×90 mm, 31,9 g
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,10 €
    16,10 €
    1 * 100 ST
       16,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    662-0242
    Shelf life
    4 Year(s)
  • 662-2339
    pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax
    pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax
    6,0 - 10,0
    Classic flat box , 65×10×90 mm, 34,36 g
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,10 €
    16,10 €
    1 * 100 ST
       16,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    662-2339
    Shelf life
    4 Year(s)
  • 662-0207
    pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax
    pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax
    7,0 - 14,0
    Classic flat box , 66×10×90 mm, 34,51 g
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,10 €
    16,10 €
    1 * 100 ST
       16,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    662-0207
    Shelf life
    4 Year(s)
  • 662-0236
    pH-Fix, 100 test strips with colour scale
    pH-Fix, 100 test strips with colour scale
    7,5 - 9,5
    Classic flat box , 66×10×90 mm, 34,58 g
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,10 €
    16,10 €
    1 * 100 ST
       16,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    662-0236
    Shelf life
    4 Year(s)
  • 662-0237
    pH-Fix, 100 test strips with colour scale
    pH-Fix, 100 test strips with colour scale
    7,9 - 9,8
    Classic flat box , 65×10×90 mm, 32 g
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,10 €
    16,10 €
    1 * 100 ST
       16,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    662-0237
    Shelf life
    4 Year(s)

Plop-Top tube container with colour chart

Product Information

  • pH range
    Pack type
    Availability
    Price
  • 662-2337
    pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax
    pH-Fix, 100 test strips with colour scale, for use with QUANTOFIX Relax
    0,0 - 14,0
    PlopTop tube , 35×103×27 mm, 25,5 g
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,10 €
    16,10 €
    1 * 100 ST
       16,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    662-2337
    Shelf life
    4 Year(s)
  • 662-2338
    pH-Fix, 100 test strips with colour scale, CE-certified according to IVDR 2017/746, for professional use
    pH-Fix, 100 test strips with colour scale, CE-certified according to IVDR 2017/746, for professional use
    4,5 - 10,0
    PlopTop tube , 30×30×100 mm, 23,9 g
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 16,10 €
    16,10 €
    1 * 100 ST
       16,10 €

    Specifications:
    More
    Specifications
    Cat. No.
    662-2338
    Shelf life
    4 Year(s)