Sulphurous acid (≥ 6,0% SO₂) in aqueous solution ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Sulphur dioxide solution
CAS Number:7782-99-2
:
Formula:H₂O₃S
MDL Number:MFCD00011335
Density:1.030
Molecular Weight:82.08 g/mol
ADR:8 II
Merck Index:14,8976
EINECS:231-973-1
UN:1833
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together