Order Entry
Austria
ContactUsLinkComponent
Crystal violet ≥90% (by anhydrous basis) ACS, Sigma-Aldrich®
Corrosive
Health
Irritant
Environment
Crystal violet ≥90% (by anhydrous basis) ACS, Sigma-Aldrich®
Certificates
undefined
Corrosive
Health
Irritant
Environment
Crystal violet ≥90% (by anhydrous basis) ACS, Sigma-Aldrich®

Specification Test Results

  • Appearance (Colour):
    Green to Very Dark Green
  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Loss on Drying:
    ≤ 7.5 %
  • Sensitivity
    as Indicator:
    Pass
  • UV Absorbance Ratio:
    0.98 - 1.90
  • Purity:
    ≥90.0 %
  • Meets ACS Requirements:
    Current ACS Specification

Specification Test Results

Frequently Bought Together