ContactUsLinkComponent
Eosin Y (yellowish), pure indicator grade
Eosin Y (yellowish), pure indicator grade
Certificates
undefined
Eosin Y (yellowish), pure indicator grade
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    Dark red-brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • UV spectrum:
    Complies
  • Loss on drying:
    ≤10 %
  • UV Lambda max:
    511 to 519 nm
  • Specific extinction E (1%/1cm):
    ≥1100 (lambda max, 0.001M Na2CO3) on dried substance