Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Eosin B
Eosin B 1 g/l in aqueous solution, Sigma-Aldrich®
Eosin B 1 g/l in aqueous solution, Sigma-Aldrich®
2853-100ML
:
SIGMA ALDRICH MICROSCOPY
:
Eosin B 1 g/l in aqueous solution, Sigma-Aldrich®
2853-100ML
:
SIGMA ALDRICH MICROSCOPY
:
Pack type:
Glass bottle
Pk:
100 mL
Seq:
2440
:
:
624.06 g/mol
:
M-Clarity™ quality level = MQ500 (
more info.
)
:
208-943-1
:
548-24-3
:
C₂₀H₆Br₂N₂Na₂O₉
:
MFCD00005041