Fuchsin (acid) for tissue, stain, Sigma-Aldrich®
:3244-88-0
:
:C₂₀H₁₇N₃Na₂O₉S₃
:MFCD00013286
:585.55 g/mol
:12,00109
:> 130 °C
:M-Clarity™ quality level = MQ200
:Room Temperature
:221-816-5
- Appearance (Form):Powder
- Solubility (Colour):Red
- Solubility (Turbidity)
4 mg + 4 ml, H₂O:Clear - Nitrogen:4.3 - 7.2 %
- Recommended Retest Period:4 years
- % Dye content
(approx. 70%):≥60 %
Frequently Bought Together