Order Entry
Austria
ContactUsLinkComponent
2-Methyl-2-nitropropane-1,3-diol 99%
2-Methyl-2-nitropropane-1,3-diol 99%
Catalog # ACRO134800050
CAS Number:  
undefined
2-Methyl-2-nitropropane-1,3-diol 99%
Catalog # ACRO134800050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to yellow Crystals
  • Infrared spectrum:
    Conforms
  • GC:
    ≥98.5 %
  • NMR (31P-NMR):
    ≥97.5 %

Specification Test Results

About this item

 :
 :135.12 g/mol
Boiling Pt:324 °C (1013 hPa)
Melting Pt:147…149 °C
EINECS:201-031-4
CAS Number:77-49-6
Formula:C₄H₉NO₄
MDL Number:MFCD00024812