
2-Methyl-2-nitropropane-1,3-diol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:77-49-6
:
Formula:C₄H₉NO₄
MDL Number:MFCD00024812
:135.12 g/mol
Boiling Pt:324 °C (1013 hPa)
Melting Pt:148…151 °C
Storage Temperature:Ambient
EINECS:201-031-4
:GHS07
Related Information
Frequently Bought Together