Order Entry
Austria
ContactUsLinkComponent
1,10-Phenanthroline hydrochloride monohydrate ≥99.5% (by anhydrous basis), GR Reag. Ph. Eur. for analysis, redox indicator, Supelco®
Toxic
Environment
1,10-Phenanthroline hydrochloride monohydrate ≥99.5% (by anhydrous basis), GR Reag. Ph. Eur. for analysis, redox indicator, Supelco®
Supplier:  Merck
Certificates
Toxic
Environment
1,10-Phenanthroline hydrochloride monohydrate ≥99.5% (by anhydrous basis), GR Reag. Ph. Eur. for analysis, redox indicator, Supelco®
Supplier:  Merck

Specification Test Results

  • Assay (alcalimetric, calculated on anhydrous substance):
    ≥ 99.5 %
  • Identity (IR-spectrum):
    passes test
  • Melting point:
    about 215 °C, with decomposition
  • Substances insoluble in water:
    passes test
  • Sulfated ash:
    ≤ 0.2 %
  • Water (according to Karl Fischer):
    7.0 - 8.5 %
  • Suitability as indicator:
    passes test
  • Sensitivity test (at Iron II ions):
    passes test

Specification Test Results

Related Information

Frequently Bought Together