Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Osteo-Bed Bone Embedding Solvent

Osteo-bed bone embedding solvent, 2.000 mL
Catalog # O8639-4X500ML
Osteo-Bed Bone Embedding Solvent
Catalog # O8639-4X500ML
CAS Number:  
Osteo-Bed Bone Embedding Solvent

Osteo-bed bone embedding solvent, 2.000 mL

CAS Number:  
Price
216,00 €
Log in for availability
1 * 2.000 mL
Product Details & Documents

For use with Osteo-bed bone embedding kit. Solvent for removal of plastic from sections prior to rehydration and staining. Most histochemical and immunohistochemistry staining protocols require the removal of methyl methacrylate (MMA) and rehydration of mounted sections for improved stain penetration and greater contrast.

  • Biological source is synthetic
Specifications

Specifications

  • Pk:
    2.000 mL
  • Description:
    Osteo-bed bone embedding solvent
  • Application:
    Hematology<br />Histology
  • Cat. No.:
    O8639-4X500ML
  • Storage temperature:
    RT
  • Density:
    >1,20 g/ml

Specifications