Rhodamine B isothiocyanate–Dextran
R8881-100MG
:
- Pk:100 mg
- Seq:2440
- Appearance (Colour)
Light pink to red to very dark purple:Conforms - Appearance (Form):Powder
- Solubility (Colour):Pink
- Solubility (Turbidity)
10 mg/mL, H2O:Clear - Degree of Substitution
Mole RITC per Mole Glucose:0.002 - 0.010 - Average Molecular Weight
of Dextran:9000 - 11000
:
:Refrigerator
:M-Clarity™ quality level = MQ200
:MFCD00132176