TISSUE-TROL™ Control Slides Helicobacter
Helicobacter TISSUE-TROL™ control slides from mouse intestine (tissue containing helicobacter pylori)
Catalog # TTR006-25EA
Supplier: SIGMA ALDRICH MICROSCOPY
Jump to:
Product Details & Documents
Histology slides with paraffin embedded section of mouse intestine tissue containing Helicobacter pylori. Suitable for use with modified steiner-steiner silver stain procedure No. HT101.
Delivery information: Box contains 24 unstained slides plus one reference slide.
Documents
Merck The Color of Tissue Diagnostics
Product Literature
Specifications
- Description:Helicobacter TISSUE-TROL™ control slides from mouse intestine (tissue containing helicobacter pylori)