Orders LinkContactUsLinkComponent
Fluid Point Sampler
Fluid Point Sampler
Supplier:   Sampling Systems
Fluid Point Sampler
Supplier:  Sampling Systems
Certificates
Fluid Point Sampler
Fluid Point Sampler
3 Options Available Below
Product Details & Documents
The fluid point sampler is designed for taking rapid samples of free-flowing liquids.
  • Ideal for sampling free-flowing liquids from most containers
  • Great for sampling pharmaceuticals, cosmetics, and foodstuffs
  • Easily cleaned
  • Available from stock
  • Fast delivery
The fluid point sampler is great for taking samples of free-flowing liquids from a range of containers, including drums. It is made from chemically resistant 316L stainless steel to provide excellent corrosion resistance.
Product Family Options
Product Information
  • Material
    Capacity
    Length
    Ø
    Availability
    Price
  • SAMP5551S-600
    Fluid Point sampler
    Fluid Point sampler
    Stainless steel 316L
    160 ml
    600 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 843,00 €
    843,00 €
    1 * 1 ST
       843,00 €
  • SAMP5551S-1000
    Fluid Point sampler
    Fluid Point sampler
    Stainless steel 316L
    300 ml
    1000 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 939,00 €
    939,00 €
    1 * 1 ST
       939,00 €
  • SAMP5551S-1500
    Fluid Point sampler
    Fluid Point sampler
    Stainless steel 316L
    450 ml
    1500 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 060,00 €
    1 060,00 €
    1 * 1 ST
       1 060,00 €