Orders LinkContactUsLinkComponent
Silo Rocket Sampler
Silo Rocket Sampler
Silo Rocket Sampler
Silo Rocket Sampler
The Silo rocket sampler is ideal for taking large bulk samples.
  • Suitable for both cohesive and free-flowing powders
  • Can be used for grains, granules, etc.
  • Fitted with a wide handle for extra leverage
  • Available from stock
  • Fast delivery
The Silo rocket sampler features a convenient design, meaning it can easily take samples of both free-flowing and cohesive powders. It is made from high-quality 316L stainless steel for durability.
Product Family Options
  • Description
    Material
    Length
    Volume
  • Silo Rocket sampler
    Stainless steel 316L
    1000 mm
    500 ml
    1 * 1 ST
    1 * 1 ST - 3 470,00 €
    3 470,00 €
    1 * 1 ST
       3 470,00 €
  • Silo Rocket sampler
    Stainless steel 316L
    1500 mm
    500 ml
    1 * 1 ST
    1 * 1 ST - 3 520,00 €
    3 520,00 €
    1 * 1 ST
       3 520,00 €
  • Silo Rocket sampler
    Stainless steel 316L
    2000 mm
    500 ml
    1 * 1 ST
    1 * 1 ST - 3 520,00 €
    3 520,00 €
    1 * 1 ST
       3 520,00 €