Orders LinkContactUsLinkComponent
Jumbo Sack Sampler
Jumbo Sack Sampler
Jumbo Sack Sampler
Jumbo Sack Sampler
The jumbo sack sampler is designed for sampling large granules from sacks.
  • Ideal for sampling granules from bags on pallets
  • Can sample larger granules up to 10mm in size
  • Simple to use
  • Ideal for most powders and granules
  • Available from stock
  • Fast delivery
The jumbo aack sampler is made from 316L Stainless steel, and is perfect for sampling powders and granules in sacks. There are various models to suit specific applications, including a model with a plain end which the user can attach to their own bag. A model with an end cap allows the user to retain the sample in the tube. Additionally, a model is available with an adaptor to enable the user to collect the sample in a bottle.
Product Family Options
  • Description
    Material
    Length
    Ø
  • Jumbo Sack sampler (plain)
    Stainless steel 316L
    400 mm
    40 mm
    1 * 1 ST
    1 * 1 ST - 272,00 €
    272,00 €
    1 * 1 ST
       272,00 €
  • Jumbo Sack sampler (plain)
    Stainless steel 316L
    800 mm
    40 mm
    1 * 1 ST
    1 * 1 ST - 322,00 €
    322,00 €
    1 * 1 ST
       322,00 €
  • Jumbo Sack sampler (with end cap)
    Stainless steel 316L
    400 mm
    40 mm
    1 * 1 ST
    1 * 1 ST - 471,00 €
    471,00 €
    1 * 1 ST
       471,00 €
  • Jumbo Sack sampler (with end cap)
    Stainless steel 316L
    800 mm
    40 mm
    1 * 1 ST
    1 * 1 ST - 543,00 €
    543,00 €
    1 * 1 ST
       543,00 €
  • Jumbo Sack sampler with 1000ml bottle
    Stainless steel 316L
    400 mm
    40 mm
    1 * 1 ST
    1 * 1 ST - 608,00 €
    608,00 €
    1 * 1 ST
       608,00 €
  • Jumbo Sack sampler with 1000ml bottle
    Stainless steel 316L
    800 mm
    40 mm
    1 * 1 ST
    1 * 1 ST - 670,00 €
    670,00 €
    1 * 1 ST
       670,00 €