Stopper, IN-Stopper
Catalog # 720-2581
Supplier: B. Braun
Specifications
- Description:IN-Stopper closure stopper with injection port, Priming volume: 0,20 ml, pressure resistance: 2 bar, shelf life: 60 months
- Environmentally Preferable:
- Pk:100
Specifications
About this item
IN-Stopper is a male luer lock closing cone with injection port for intermittent injections through injection membrane. It closes female accesses and offers a needle access via an injection membrane, e.g. to an infusion solution and has got a smooth, hygienic outer surface.
- Smooth, hygienic outer surface
- Offers a needle access via a membrane, which is not made with Latex
- IN-Stopper can be used with alcohol and lipids
- IN-Stopper are non-toxic and non-pyrogenic
- Easy and convenient in handling
- Five insertions possible
The device can be used for all patients for which infusion therapy is prescribed. No gender or age-related limitations. IN-Stopper can be used for adults, pediatric and neonates.