Order Entry
Austria
ContactUsLinkComponent
Chemical Oxygen Demand COD 5000 mg/L Calibration Standard
Chemical Oxygen Demand COD 5000 mg/L Calibration Standard
Catalog # ACSDCOD5000.L5
Supplier:  CPACHEM
New Product
CAS Number:  
Chemical Oxygen Demand COD 5000 mg/L Calibration Standard
Catalog # ACSDCOD5000.L5
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Standards:
    Calibration standard
  • Description:
    Chemical oxygen demand COD 5000 mg/L calibration standard
  • COD:
    5000 mg/L

Specifications

About this item

Shelf life on ship date: 24 months - store in a refrigerator at temperatures between 2 to 8 °C.