Total Nitrogen Standard (1000 mg/L) - for Photometric Applications
Catalog # ACSDTN1000.L1
Supplier: CPACHEM
CAS Number:
Specifications
- Pk:100 mL
- Description:Total nitrogen standard (1000 mg/L) - for photometric applications
- Shelf life:24 months
- Storage temperature:15...25 °C