Order Entry
Austria
ContactUsLinkComponent
Chromium ICP and ICP-MS CRM, Chromium (III) 1000 mg/L in Diluted Nitric Acid
Chromium ICP and ICP-MS CRM, Chromium (III) 1000 mg/L in Diluted Nitric Acid
Artikelnr. ACSDSP073.2N.L1
Supplier:  CPACHEM
CAS Number:  
Chromium ICP and ICP-MS CRM, Chromium (III) 1000 mg/L in Diluted Nitric Acid
Artikelnr. ACSDSP073.2N.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Pk:
    100 mL
  • CAS No.:
    7697-37-2
  • Description:
    Chromium (III) 1000 mg/L in diluted nitric acid
  • Hazard class description:
    8 Corrosive substances
  • UN No.:
    1760
  • GHS signal:
    Warning

Specifications

About this item

Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.